missing translation for 'onlineSavingsMsg'
Learn More

Survivin Ab-6, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Survivin Ab-6, Rabbit Polyclonal Antibody.

£133.62 - £617.10

Specifications

Antigen Survivin Ab-6
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Formulation Price Quantity & Availability  
Product Code Brand Quantity Formulation Price Quantity & Availability  
12603087
View Documents
Epredia
RB-1629-P0
100 μL Purified with BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12613087
View Documents
Epredia
RB-1629-P1
500 μL Purified with BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12623087
View Documents
Epredia
RB-1629-P1ABX
500 μL Purified w/o BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12693077
View Documents
Epredia
RB-1629-P
1 mL Purified with BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12633087
View Documents
Epredia
RB-1629-PABX
1 mL Purified w/o BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Survivin, a new inhibitor of apoptosis (IAP) protein, is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. It indicates that survivin may counteract a default induction of apoptosis in G2/M phase. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favour aberrant progression of transformed cells through mitosis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of survivin

Molecular Weight: 16.5kDa

Positive Control: Stomach

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Survivin Ab-6
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis
A synthetic peptide derived from the C-terminus of survivin
Primary
1mg/mL
Polyclonal
Concentrated
RUO
Human
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.