missing translation for 'onlineSavingsMsg'
Learn More

Survivin Ab-6, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Survivin Ab-6, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-1629-P

 View more versions of this product

Product Code. 12693077

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Survivin, a new inhibitor of apoptosis (IAP) protein, is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. It indicates that survivin may counteract a default induction of apoptosis in G2/M phase. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favour aberrant progression of transformed cells through mitosis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of survivin

Molecular Weight: 16.5kDa

Positive Control: Stomach

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Survivin Ab-6
Polyclonal
1mg/mL
Purified with BSA and Azide
A synthetic peptide derived from the C-terminus of survivin
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
Rabbit
1 mL
Apoptosis
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.