missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 Mouse anti-Human, Mouse, Rat, CoraLite 647, Clone: 3G2D12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | STAT3 |
|---|---|
| Clone | 3G2D12 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17711469
|
Proteintech
CL647-60199-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17701469
|
Proteintech
CL647-60199-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for FC (Intra)Specifications
| STAT3 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| 6774 | |
| Primary | |
| Store at -20°C |
| 3G2D12 | |
| Flow Cytometry | |
| CoraLite 647 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IgG2a | |
| Protein A purification | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title