missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 Mouse anti-Human, Mouse, Rat, CoraLite 647, Clone: 3G2D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-60199-20UL
This item is not returnable.
View return policy
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for FC (Intra)Specifications
| STAT3 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Mouse | |
| 20 μL | |
| 6774 | |
| Store at -20°C | |
| Liquid |
| Flow Cytometry | |
| 3G2D12 | |
| CoraLite 647 | |
| BC000163 | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction