missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta III Mouse, Clone: SPTBN2/1584, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Spectrin beta III |
---|---|
Clone | SPTBN2/1584 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950918
|
EnquireBio
6712-MSM4-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940918
|
EnquireBio
6712-MSM4-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Spectrin beta III | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
6712 | |
Recombinant human SPTBN2 fragment (aa356-475) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
246kDa |
SPTBN2/1584 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_016873664 | |
SPTBN2 | |
IgG1 | |
Protein A/G Purified | |
Cell Surface | |
HeLa cells. Pancreas or Liver. |