missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta III Mouse, Clone: SPTBN2/1584, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6712-MSM4-P0
This item is not returnable.
View return policy
Specifications
| Spectrin beta III | |
| Cell Surface | |
| SPTBN2/1584 | |
| Unconjugated | |
| XP_016873664 | |
| Mouse | |
| 246kDa | |
| Protein A/G Purified | |
| RUO | |
| 6712 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| SPTBN2 | |
| Recombinant human SPTBN2 fragment (aa356-475) | |
| HeLa cells. Pancreas or Liver. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction