Learn More
Sodium nitrate, 98+%, Thermo Scientific™
£19.40 - £198.00
Chemical Identifiers
| CAS | 7631-99-4 |
|---|---|
| Molecular Formula | NNaO3 |
| Molecular Weight (g/mol) | 84.994 |
| MDL Number | MFCD00011119 |
| InChI Key | VWDWKYIASSYTQR-UHFFFAOYSA-N |
| Synonym | sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 |
| PubChem CID | 24268 |
| ChEBI | CHEBI:63005 |
| IUPAC Name | sodium;nitrate |
| SMILES | [N+](=O)([O-])[O-].[Na+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11449443
|
Thermo Scientific Alfa Aesar
A12327.0C |
10,000g |
£198.00
10000g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11429443
|
Thermo Scientific Alfa Aesar
A12327.36 |
500g |
£19.40
500g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11439443
|
Thermo Scientific Alfa Aesar
A12327.0E |
2500g |
£57.60 £51.65 / 2500g Save £5.95 10% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsSodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.
Solubility
Freely soluble in water, ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.
Notes
Sodium nitrate is hygroscopic in nature.
Chemical Identifiers
| NNaO3 | |
| MFCD00011119 | |
| sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
| CHEBI:63005 | |
| [N+](=O)([O-])[O-].[Na+] |
Specifications
| 7631-99-4 | |
| MFCD00011119 | |
| 14,8647 | |
| Freely soluble in water,ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine. | |
| [N+](=O)([O-])[O-].[Na+] | |
| 84.994 | |
| CHEBI:63005 | |
| ≥98% | |
| Hygroscopic | |
| Sodium nitrate |
| NNaO3 | |
| UN1498 | |
| sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
| VWDWKYIASSYTQR-UHFFFAOYSA-N | |
| sodium;nitrate | |
| 24268 | |
| 84.99 | |
| Odorless | |
| 2.26 |
Safety and Handling
GHS H Statement
H272-H302
May intensify fire
oxidizer.
Harmful if swallowed.
P210-P220-P221-P260-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H272-H319-H373
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: SODIUM NITRATE
missing translation for 'einecsNumber' : 231-554-3
missing translation for 'rtecsNumber' : WC5600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium nitrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contain: Sodium nitrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.