missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, 98+%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar A12327.36


Quantity 500g

 View more versions of this product

Product Code. 11429443

  • £19.40 / 500g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.

Solubility
Freely soluble in water, ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.

Notes
Sodium nitrate is hygroscopic in nature.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7631-99-4
84.994
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
sodium;nitrate
NNaO3
MFCD00011119
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
CHEBI:63005
[N+](=O)([O-])[O-].[Na+]

Specifications

7631-99-4
NNaO3
UN1498
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium;nitrate
24268
84.99
Odorless
2.26
500g
MFCD00011119
14,8647
Freely soluble in water,ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.
[N+](=O)([O-])[O-].[Na+]
84.994
CHEBI:63005
≥98%
Hygroscopic
Sodium nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium nitrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contain: Sodium nitrate

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.