missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, 98+%, Thermo Scientific™

Marca:  Thermo Scientific Alfa Aesar A12327.0E


Quantity 2500g

 Ver más versiones de este producto

Código de producto. 11439443

  • ONLINE EXCLUSIVE PRICE
    £57.60
    £51.65 / 2500g
    Save £5.95
    10% Off
 Restricted Item
In Stock
Añadir a la cesta
Request Bulk or Custom Formats
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.

Solubility
Freely soluble in water, ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.

Notes
Sodium nitrate is hygroscopic in nature.
TRUSTED_SUSTAINABILITY
Especificaciones

Identificadores de productos químicos

7631-99-4
84.994
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
sodium;nitrate
NNaO3
MFCD00011119
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
CHEBI:63005
[N+](=O)([O-])[O-].[Na+]

Especificaciones

7631-99-4
NNaO3
UN1498
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium;nitrate
24268
84.99
Odorless
2.26
2500g
MFCD00011119
14,8647
Freely soluble in water,ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.
[N+](=O)([O-])[O-].[Na+]
84.994
CHEBI:63005
≥98%
Hygroscopic
Sodium nitrate
Sugerencias de productos

Sugerencias de productos

Videos
Seguridad y manipulación

Seguridad y manipulación

Nombre del producto
  • Sodium nitrate

Palabra de advertencia
  • Warning

Clasificación
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3

Indicaciones de peligro
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.

Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

Otros peligros
  • MIXTURE LIST-Contain: Sodium nitrate

SDS
Documentos

Documentos

Product Certifications
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.