Learn More
Description
Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.
- Reactive groups: NHS ester and maleimide
- Reactive toward: amino and sulfhydryl groups
- Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
- Polyethylene glycol spacer arms help maintain conjugate solubility
- Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Ideal for performing controlled, two-step protein conjugations
Specifications
Specifications
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | No |
| Description | BS(PEG)5 |
| Molecular Weight (g/mol) | 532.5 |
| PEGylated | Yes |
| Spacer Arm Length | 53.4 Å |
| Content And Storage | Upon receipt store desiccated at -20°C. |
| Cell Permeability | No |
| Shipping Condition | Wet Ice |
| Product Line | No-Weigh, Pierce |
| Show More |
Frequently Asked Questions (FAQs)
SM(PEG)12 (PEGylated, long-chain SMCC crosslinker) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Sometimes. The biotin has to insert 9 angstrom into the pocket of avidin, streptavidin or NeutrAvidin to bind. If steric hindrance may be a problem, using a biotinylation reagent with a longer spacer arm (Sulfo-NHS-LC Biotin or Sulfo-NHS-LC-LC Biotin) may result in greater purification or deteciton.
The sodium sulfonate group lends water solubility to the reagent.
Note: The actual biotin moiety that is added is identical between NHS and Sulfo-NHS biotins that vary only in the addition of the sodium sulfonate on the leaving group. Researchers preferring a more water soluble biotin be added to their molecule should consider a PEGylated version of biotin.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.