missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC44A4 Antibody (ASG-5ME) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | SLC44A4 |
|---|---|
| Clone | ASG-5ME |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046891
|
Novus Biologicals™
NBP3-28624-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045430
|
Novus Biologicals™
NBP3-28624-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044323
|
Novus Biologicals™
NBP3-28624-100ug |
100 μg |
£266.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC44A4 Monoclonal antibody specifically detects SLC44A4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| SLC44A4 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Endocrinology, Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C6orf29, Choline Transporter-Like Protein 4, Chromosome 6 Open Reading Frame 29, CTL4, NG22, Solute Carrier Family 44 Member 4, Solute Carrier Family 44, Member 4 | |
| SLC44A4 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| ASG-5ME | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q53GD3 | |
| 80736 | |
| IgG2 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title