missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC44A4 Antibody (ASG-5ME) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28624-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SLC44A4 Monoclonal antibody specifically detects SLC44A4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Especificaciones
| SLC44A4 | |
| Monoclonal | |
| Unconjugated | |
| Q53GD3 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| ASG-5ME | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C6orf29, Choline Transporter-Like Protein 4, Chromosome 6 Open Reading Frame 29, CTL4, NG22, Solute Carrier Family 44 Member 4, Solute Carrier Family 44, Member 4 | |
| SLC44A4 | |
| 100 μg | |
| Endocrinology, Signal Transduction | |
| 80736 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido