missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC44A4 Antibody (ASG-5ME) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28624-50ug
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
SLC44A4 Monoclonal antibody specifically detects SLC44A4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifica
| SLC44A4 | |
| Monoclonal | |
| Unconjugated | |
| Q53GD3 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| ASG-5ME | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C6orf29, Choline Transporter-Like Protein 4, Chromosome 6 Open Reading Frame 29, CTL4, NG22, Solute Carrier Family 44 Member 4, Solute Carrier Family 44, Member 4 | |
| SLC44A4 | |
| 50 μg | |
| Endocrinology, Signal Transduction | |
| 80736 | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto