missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC44A3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | SLC44A3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SLC44A3 Polyclonal antibody specifically detects SLC44A3 in Human samples. It is validated for Western BlotSpecifications
| SLC44A3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 126969 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| choline transporter-like protein 3, member 3, MGC45474, solute carrier family 44, member 3, UNQ558/PRO1115 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-220 of human SLC44A3 (NP_001107578.1). RLLFGYDSFGNMCGKKNSPVEGAPLSGQDMTLKKHVFFMNSCNLEVKGTQLNRMALCVSNCPEEQLDSLEEVQFFANTSGSFLCVYSLNSFNYTHSPKADSLCPRLPVPPSKSFPLFNRCVPQTPECYSLFASVLINDVDTLHRILSGIMSGRDTILGLCI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title