missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC44A3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93370-0.1ml
This item is not returnable.
View return policy
Description
SLC44A3 Polyclonal antibody specifically detects SLC44A3 in Human samples. It is validated for Western Blot
Specifications
| SLC44A3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| choline transporter-like protein 3, member 3, MGC45474, solute carrier family 44, member 3, UNQ558/PRO1115 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-220 of human SLC44A3 (NP_001107578.1). RLLFGYDSFGNMCGKKNSPVEGAPLSGQDMTLKKHVFFMNSCNLEVKGTQLNRMALCVSNCPEEQLDSLEEVQFFANTSGSFLCVYSLNSFNYTHSPKADSLCPRLPVPPSKSFPLFNRCVPQTPECYSLFASVLINDVDTLHRILSGIMSGRDTILGLCI | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 126969 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering