missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SLC38A3 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16872894
|
Proteintech
14315-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16862894
|
Proteintech
14315-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.Specifications
| SLC38A3 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| Q99624, Q9DCP2, Q9JHZ9 | |
| 10991, 252919, 76257 | |
| SLC38A3 Fusion Protein Ag5553 | |
| Primary | |
| -20°C | |
| SLC38A3 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| G17, NAT1, SLC38A3, SN1, SNAT3 | |
| Slc38a3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title