missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14315-1-AP-20UL
This item is not returnable.
View return policy
Description
As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.Specifications
| SLC38A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A3 | |
| G17, NAT1, SLC38A3, SN1, SNAT3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10991, 252919, 76257 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot, Flow Cytometry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99624, Q9DCP2, Q9JHZ9 | |
| Slc38a3 | |
| SLC38A3 Fusion Protein Ag5553 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction