missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14315-1-AP-150UL
This item is not returnable.
View return policy
Beskrivning
As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.Specifikationer
| SLC38A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A3 | |
| G17, NAT1, SLC38A3, SN1, SNAT3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10991, 252919, 76257 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99624, Q9DCP2, Q9JHZ9 | |
| Slc38a3 | |
| SLC38A3 Fusion Protein Ag5553 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering