missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Silicone Sealing Ring, with bonded PTFE face

Product Code. 11797118 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inside:
11.0 to 13.0 mm
9.0 to 11.0 mm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Unit Size
11797118 11.0 to 13.0 mm Pack of 10
11767118 9.0 to 11.0 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11797118 Supplier DWK Life Sciences Supplier No. 292361201
  • ONLINE EXCLUSIVE PRICE
    £118.80
    £96.21 / Pack of 10
    Save £22.59
    19% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The silicone seal is used for sealing and fixing threaded pipes with threaded caps.

The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.

  • With bonded PTFE face

Specifications

Diameter (Metric) Inside 11.0 to 13.0 mm
For Use With (Application) Sealing; Connecting glass tubes
Joint Size GL 32
Quantity 10/Pk.
Size (Metric) Fits Tubing ID: 11.0 to 13.0 mm
Diameter (Metric) Outside 29 mm
Material Silicone (VMQ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.