Learn More
Duran™ Silicone and PTFE Sealing Rings
Product Code. 15814628 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size (Metric):
S 13
S 19
S 35
S 41
S 64
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) unitSize
15814628 S 13 Pack of 10
15824628 S 19 Pack of 10
15834628 S 35 Pack of 10
15961561 S 41 Pack of 10
15971561 S 64 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15814628 Supplier Duran™ Supplier No. 286222202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
For Use With (Application) For Polished Spherical Joints
Quantity 10 Pack
Size (Metric) S 13
Material PTFE, Silicone
Type Seal Ring