missing translation for 'onlineSavingsMsg'
Learn More

Sheep IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE

Donkey Polyclonal Antibody

£271.00 - £2170.00

Specifications

Antigen IgG
Dilution Immunohistochemistry
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP Polymer
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18785808
GREENER_CHOICE
View Documents
R&D Systems
VC006-025
25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18723579
GREENER_CHOICE
View Documents
R&D Systems
VC006-050
50 mL (2 x 25 mL)
£433.00
50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18784098
GREENER_CHOICE
View Documents
R&D Systems
VC006-125
125 mL
£1140.00
125mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18717828
GREENER_CHOICE
View Documents
R&D Systems
VC006-250
250 mL (2 x 125 mL)
£2170.00
250mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Immunohistochemistry.
Specifications

Specifications

IgG
Immunohistochemistry
HRP Polymer
RUO
Supplied as a solution in PBS with preservative.
3500
IgG
Affinity Purified
Detects heavy ( gamma ) chains on sheep IgG and light chains on all sheep immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin sheep serum proteins or IgG from human, mouse or rabbit, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
Immunohistochemistry
Polyclonal
Donkey
Sheep
Immunoglobulin G, ImmunoglobulinG
Sheep IgG
Secondary
6 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications

RUO

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.