missing translation for 'onlineSavingsMsg'
Learn More

Sheep IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18784098
Change view
Click to view available options
Quantity:
125 mL
25 mL
250 mL (2 x 125 mL)
50 mL (2 x 25 mL)
Unit Size:
125mL
250mL
25mL
50mL
Product Code. Quantity unitSize
18784098 125 mL 125mL
18785808 25 mL 25mL
18723579 50 mL (2 x 25 mL) 50mL
18717828 250 mL (2 x 125 mL) 250mL
4 options
This item is not returnable. View return policy

Product Code. 18784098

Brand: R&D Systems VC006125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP Polymer
Dilution Immunohistochemistry
Formulation Supplied as a solution in PBS with preservative.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Donkey
Immunogen Sheep IgG
Purification Method Affinity Purified
Quantity 125 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects heavy ( gamma ) chains on sheep IgG and light chains on all sheep immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin sheep serum proteins or IgG from human, mouse or rabbit, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
Target Species Sheep
Content And Storage 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.