missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Description
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Immunohistochemistry.
Specifications
Specifications
| Antigen | IgG |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | HRP Polymer |
| Dilution | Immunohistochemistry |
| Formulation | Supplied as a solution in PBS with preservative. |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Host Species | Donkey |
| Immunogen | Sheep IgG |
| Purification Method | Affinity Purified |
| Show More |
RUO
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?