missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGLT2/SLC5A2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £378.00
Specifications
| Antigen | SGLT2/SLC5A2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688920
|
Novus Biologicals
NBP2-77441 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604819
|
Novus Biologicals
NBP2-77441SS |
0.025 mg |
£167.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SGLT2/SLC5A2 Polyclonal antibody specifically detects SGLT2/SLC5A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SGLT2/SLC5A2 | |
| Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 2, SGLT2sodium/glucose cotransporter 2, solute carrier family 5 (sodium/glucose cotransporter), member 2, solute carrier family 5 (sodium/glucose transporter), member 2, Solute carrier family 5 member 2 | |
| Partial synthetic peptide made to an internal portion of the human SLC5A2 protein (between amino acids 480-540) [UniProt P31639] | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS | |
| 6524 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title