missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGLT2/SLC5A2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-77441SS
This item is not returnable.
View return policy
Description
SGLT2/SLC5A2 Polyclonal antibody specifically detects SGLT2/SLC5A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SGLT2/SLC5A2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| Low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 2, SGLT2sodium/glucose cotransporter 2, solute carrier family 5 (sodium/glucose cotransporter), member 2, solute carrier family 5 (sodium/glucose transporter), member 2, Solute carrier family 5 member 2 | |
| Partial synthetic peptide made to an internal portion of the human SLC5A2 protein (between amino acids 480-540) [UniProt P31639] | |
| 0.025 mg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 6524 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction