missing translation for 'onlineSavingsMsg'
Learn More

SGLT2/SLC5A2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-77441

 View more versions of this product

Product Code. 18688920

  • £378.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SGLT2/SLC5A2 Polyclonal antibody specifically detects SGLT2/SLC5A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SGLT2/SLC5A2
Polyclonal
Unconjugated
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200
Low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 2, SGLT2sodium/glucose cotransporter 2, solute carrier family 5 (sodium/glucose cotransporter), member 2, solute carrier family 5 (sodium/glucose transporter), member 2, Solute carrier family 5 member 2
Partial synthetic peptide made to an internal portion of the human SLC5A2 protein (between amino acids 480-540) [UniProt P31639]
0.1 mg
Apoptosis, Cancer, Tumor Suppressors
6524
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.