Learn More
DWK Life Sciences Sealing Ring, for polished spherical joints (VITON)
Product Code. 15824498 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
13 NS
19 NS
29 NS
64 NS
Unit Size:
Pack of 10
Product Code. 15824498
Brand: DWK Life Sciences 1155357
Only null left
Add to basket
Add to basket
For sealing ground joints without ground joints DURAN™, especially for vacuum applications.
Ball joints without ground joints only seal via the sealing ring. VITON sealing rings are used in vacuum applications.
  • Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
  • Sealing ring made of VITON
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Sealing of non-sanded spherical ground joints
Joint Size 64 NS
Quantity 10/Pk.
Material Viton