Learn More
DWK Life Sciences Sealing Ring, for polished spherical joints (VITON)
Product Code. 15804498 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
13 NS
19 NS
29 NS
64 NS
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Size unitSize
15804498 13 NS Pack of 10
15814498 19 NS Pack of 10
15824498 64 NS Pack of 10
15894488 29 NS Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15804498 Supplier DWK Life Sciences Supplier No. 1155355
  • ONLINE EXCLUSIVE PRICE
    £41.91
    £41.14 / Pack of 10
    Save £0.77
    1% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
For sealing ground joints without ground joints DURAN™, especially for vacuum applications.
Ball joints without ground joints only seal via the sealing ring. VITON sealing rings are used in vacuum applications.
  • Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
  • Sealing ring made of VITON
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Sealing of non-sanded spherical ground joints
Joint Size 13 NS
Quantity 10/Pk.
Material Viton