missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RED Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | RED |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635540
|
Novus Biologicals
NBP2-94615-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615611
|
Novus Biologicals
NBP2-94615-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RED Polyclonal antibody specifically detects RED in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RED | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research | |
| PBS (pH 7.3), 50% glycerol | |
| 3550 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| chondrosarcoma-associated protein 2, CSA2, Cytokine IK, IK cytokine, down-regulator of HLA II, IK factor, MGC59741, protein Red, Protein RER, RD element, REDprer protein, RER | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human IK (NP_006074.2). MPERDSEPFSNPLAPDGHDVDDPHSFHQSKLTNEDFRKLLMTPRAAPTSAPPSKSRHHEMPREYNEDEDPAARRRKKKSYYAKLRQQEIERERELAEKYRDRAKERRDGVNKDYEETELISTTANYRAVGPTAEADKSAAEKRRQLIQESKFLGGDMEHTHLVKGLDFALLQKVRAEIASKEKEEEELMEKPQKETKKDEDPENKIEFKTRLGRNVYRMLFKSKAYERNELFLPGRMAYVVDLDDEYADTDIPTTLIRSKADCPTMEAQTTLTTNDIVIS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title