missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RED Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94615-0.1ml
This item is not returnable.
View return policy
Description
RED Polyclonal antibody specifically detects RED in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RED | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| chondrosarcoma-associated protein 2, CSA2, Cytokine IK, IK cytokine, down-regulator of HLA II, IK factor, MGC59741, protein Red, Protein RER, RD element, REDprer protein, RER | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human IK (NP_006074.2). MPERDSEPFSNPLAPDGHDVDDPHSFHQSKLTNEDFRKLLMTPRAAPTSAPPSKSRHHEMPREYNEDEDPAARRRKKKSYYAKLRQQEIERERELAEKYRDRAKERRDGVNKDYEETELISTTANYRAVGPTAEADKSAAEKRRQLIQESKFLGGDMEHTHLVKGLDFALLQKVRAEIASKEKEEEELMEKPQKETKKDEDPENKIEFKTRLGRNVYRMLFKSKAYERNELFLPGRMAYVVDLDDEYADTDIPTTLIRSKADCPTMEAQTTLTTNDIVIS | |
| 0.1 mL | |
| Cytokine Research | |
| 3550 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction