missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RED Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94615-0.02ml
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
RED Polyclonal antibody specifically detects RED in Human, Mouse, Rat samples. It is validated for Western Blot
Tekniske data
| RED | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| chondrosarcoma-associated protein 2, CSA2, Cytokine IK, IK cytokine, down-regulator of HLA II, IK factor, MGC59741, protein Red, Protein RER, RD element, REDprer protein, RER | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human IK (NP_006074.2). MPERDSEPFSNPLAPDGHDVDDPHSFHQSKLTNEDFRKLLMTPRAAPTSAPPSKSRHHEMPREYNEDEDPAARRRKKKSYYAKLRQQEIERERELAEKYRDRAKERRDGVNKDYEETELISTTANYRAVGPTAEADKSAAEKRRQLIQESKFLGGDMEHTHLVKGLDFALLQKVRAEIASKEKEEEELMEKPQKETKKDEDPENKIEFKTRLGRNVYRMLFKSKAYERNELFLPGRMAYVVDLDDEYADTDIPTTLIRSKADCPTMEAQTTLTTNDIVIS | |
| 0.02 mL | |
| Cytokine Research | |
| 3550 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion