missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Fetuin-B / FETUB Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Fetuin-B / FETUB, was expressed in the hosts and tags indicated.
£261.83 - £2888.42
Specifications
Accession Number | NP_445800.2 |
---|---|
Gene ID (Entrez) | 83928 |
Name | Fetuin-B / FETUB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048951
|
enQuireBio™
QP9897-YE-100UG |
100 μg |
£670.00
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088951
|
enQuireBio™
QP9897-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058951
|
enQuireBio™
QP9897-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078951
|
enQuireBio™
QP9897-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098951
|
enQuireBio™
QP9897-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068951
|
enQuireBio™
QP9897-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_445800.2 | |
Fetuin-B / FETUB Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
83928 | |
Research Use Only | |
Fetub | |
Recombinant Protein | |
Yeast | |
RSPPAPPLPN APFAPLRPLG CNDSEVLAVA GFALQNINRV QKDGYMLTLN RVHDARVHRQ EDMGSLFYLM LDVLETGCHV LSRKALKDCG PRIFYETVHG QCKAMFHVNK PRRVLYLPAY NCTLRPVSKR KIHSMCPDCP HPVDLSAPSV LEAATESLAK FNSENPSKQY ALVKVTKATT QWVVGPSYFV EYLIKESPCT QSQDSCSLQA SDSEPVGLCQ GSLIKSPGVP PQRFKKTVTV SCEFFESQDQ VPGGENPADT QDAKKLPQKN TAPTSSPSIT APRGSIQHLP EQEEPEDSKG KSPEEPFPVQ LDLTTNPQGD TLDVSFLYLE PEEKKLVVLP FPGKEQRSPE CPGPEKQRTP | |
Greater than 90% as determined by SDS-PAGE. |