missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Fetuin-B / FETUB Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Fetuin-B / FETUB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9897-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088951

  • £1930.00 / 500µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_445800.2
Fetuin-B / FETUB Protein
Research Use Only
Fetub
Recombinant Protein
Yeast
RSPPAPPLPN APFAPLRPLG CNDSEVLAVA GFALQNINRV QKDGYMLTLN RVHDARVHRQ EDMGSLFYLM LDVLETGCHV LSRKALKDCG PRIFYETVHG QCKAMFHVNK PRRVLYLPAY NCTLRPVSKR KIHSMCPDCP HPVDLSAPSV LEAATESLAK FNSENPSKQY ALVKVTKATT QWVVGPSYFV EYLIKESPCT QSQDSCSLQA SDSEPVGLCQ GSLIKSPGVP PQRFKKTVTV SCEFFESQDQ VPGGENPADT QDAKKLPQKN TAPTSSPSIT APRGSIQHLP EQEEPEDSKG KSPEEPFPVQ LDLTTNPQGD TLDVSFLYLE PEEKKLVVLP FPGKEQRSPE CPGPEKQRTP
Greater than 90% as determined by SDS-PAGE.
83928
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers