missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Porin P Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Porin P, was expressed in the hosts and tags indicated. option, ab
£281.57 - £2480.00
Specifications
Accession Number | NP_251969.1 |
---|---|
Gene ID (Entrez) | 882442 |
Name | P. aeruginosa Porin P Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028450
|
enQuireBio™
QP7362-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048450
|
enQuireBio™
QP7362-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038450
|
enQuireBio™
QP7362-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058450
|
enQuireBio™
QP7362-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068450
|
enQuireBio™
QP7362-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078450
|
enQuireBio™
QP7362-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_251969.1 | |
P. aeruginosa Porin P Protein | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
882442 | |
Research Use Only | |
oprP | |
Recombinant Protein | |
E. coli | |
GTVTTDGADI VIKTKGGLEV ATTDKEFSFK LGGRLQADYG RFDGYYTNNG NTADAAYFRR AYLEFGGTAY RDWKYQINYD LSRNVGNDSA GYFDEASVTY TGFNPVNLKF GRFYTDFGLE KATSSKWVTA LERNLTYDIA DWVNDNVGTG IQASSVVGGM AFLSGSVFSE NNNDTDGDSV KRYNLRGVFA PLHEPGNVVH LGLQYAYRDL EDSAVDTRIR PRMGMRGVST NGGNDAGSNG NRGLFGGSSA VEGLWKDDSV WGLEGAWALG AFSAQAEYLR RTVKAERDRE DLKASGYYAQ LAYTLTGEPR LYKLDGAKFD TIKPENKEIG AWELFYRYDS IKVEDDNIVV DSATREVGDA KGKTHTLGVN WYANEAVKVS ANYVKAKTDK ISNANGDDSG DGLVMRLQYV F | |
Greater than 90% as determined by SDS-PAGE. |