missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa Porin P Protein

A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Porin P, was expressed in the hosts and tags indicated. option, ab

Brand:  enQuireBio™ QP7362-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048450

  • £2480.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_251969.1
P. aeruginosa Porin P Protein
Research Use Only
oprP
Recombinant Protein
E. coli
GTVTTDGADI VIKTKGGLEV ATTDKEFSFK LGGRLQADYG RFDGYYTNNG NTADAAYFRR AYLEFGGTAY RDWKYQINYD LSRNVGNDSA GYFDEASVTY TGFNPVNLKF GRFYTDFGLE KATSSKWVTA LERNLTYDIA DWVNDNVGTG IQASSVVGGM AFLSGSVFSE NNNDTDGDSV KRYNLRGVFA PLHEPGNVVH LGLQYAYRDL EDSAVDTRIR PRMGMRGVST NGGNDAGSNG NRGLFGGSSA VEGLWKDDSV WGLEGAWALG AFSAQAEYLR RTVKAERDRE DLKASGYYAQ LAYTLTGEPR LYKLDGAKFD TIKPENKEIG AWELFYRYDS IKVEDDNIVV DSATREVGDA KGKTHTLGVN WYANEAVKVS ANYVKAKTDK ISNANGDDSG DGLVMRLQYV F
Greater than 90% as determined by SDS-PAGE.
882442
1 mg
Not determined.
Full Length
P. aeruginosa
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers