missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lipase A Protein
A cDNA sequence encoding the Lipase A was constructed and used to recombinantly synthesize the protein.
£99.15 - £770.00
Specifications
Name | Lipase A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | 580 units/mg powder. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955118
|
enQuireBio™
QP10772-10MG |
10 mg |
£99.15
10mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965118
|
enQuireBio™
QP10772-1GR |
1 g |
£770.00
1g |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945118
|
enQuireBio™
QP10772-100MG |
100 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Lipase A Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized without additives. |
Research Use Only | |
580 units/mg powder. | |
Other | |
Untagged | |
Greater than 90% as determined by SDS-PAGE. |