missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lipase A Protein
A cDNA sequence encoding the Lipase A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10772-10mg
Additional Details : Weight : 0.01000kg
Specifications
Lipase A Protein | |
Research Use Only | |
580 units/mg powder. | |
Other | |
Untagged | |
Greater than 90% as determined by SDS-PAGE. |
10 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized without additives. |