missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TSC22 domain family protein 1
A DNA sequence encoding the Mus musculus (Mouse) TSC22 domain family protein 4, was expressed in the hosts and tags indicated.
£261.83 - £2888.42
Specifications
Accession Number | NP_076399.4 |
---|---|
Gene ID (Entrez) | 78829 |
Name | TSC22 domain family protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009021
|
enQuireBio™
QP9922-YE-100UG |
100 μg |
£670.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049021
|
enQuireBio™
QP9922-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019021
|
enQuireBio™
QP9922-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039021
|
enQuireBio™
QP9922-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059021
|
enQuireBio™
QP9922-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029021
|
enQuireBio™
QP9922-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_076399.4 | |
TSC22 domain family protein 1 | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
78829 | |
Research Use Only | |
Tsc22d4 | |
Recombinant Protein | |
Yeast | |
MSGGKKKSSF QITSVTTDYE GPGSPGASDS PVPPALAGPP PRLPNGDPNP DPGGRGTPRN GSPPPGAPAS RFRVVKLPQG LGEPYRRGRW TCVDVYERDL EPPSFGRLLE GIRGASGGTG GRSLDSRLEL ASLGISTPIP QPGLSQGPTS WLRPPPTSPG PQARSFTGGL GQLAGPGKAK VETPPLSASP PQQRPPGPGT GDSAQTLPSL RVEVESGGSA AATPPLSRRR DGAVRLRMEL VAPAETGKVP PTDSRPNSPA LYFDASLVHK SPDPFGAAAA QSLSLARSML AISGHLDSDD DSGSGSLVGI DNKIEQAMDL VKSHLMFAVR EEVEVLKEQI RDLAERNAAL EQENGLLRAL ASPEQLAQLP SSGLPRLGPS APNGPSI | |
Greater than 90% as determined by SDS-PAGE. |