missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TSC22 domain family protein 1
A DNA sequence encoding the Mus musculus (Mouse) TSC22 domain family protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9922-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_076399.4 | |
TSC22 domain family protein 1 | |
Research Use Only | |
Tsc22d4 | |
Recombinant Protein | |
Yeast | |
MSGGKKKSSF QITSVTTDYE GPGSPGASDS PVPPALAGPP PRLPNGDPNP DPGGRGTPRN GSPPPGAPAS RFRVVKLPQG LGEPYRRGRW TCVDVYERDL EPPSFGRLLE GIRGASGGTG GRSLDSRLEL ASLGISTPIP QPGLSQGPTS WLRPPPTSPG PQARSFTGGL GQLAGPGKAK VETPPLSASP PQQRPPGPGT GDSAQTLPSL RVEVESGGSA AATPPLSRRR DGAVRLRMEL VAPAETGKVP PTDSRPNSPA LYFDASLVHK SPDPFGAAAA QSLSLARSML AISGHLDSDD DSGSGSLVGI DNKIEQAMDL VKSHLMFAVR EEVEVLKEQI RDLAERNAAL EQENGLLRAL ASPEQLAQLP SSGLPRLGPS APNGPSI | |
Greater than 90% as determined by SDS-PAGE. |
78829 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |