missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serum response factor Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum response factor, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_065239.1 |
---|---|
Gene ID (Entrez) | 20807 |
Name | Serum response factor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093081
|
enQuireBio™
QP8177-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013091
|
enQuireBio™
QP8177-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003091
|
enQuireBio™
QP8177-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023091
|
enQuireBio™
QP8177-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033091
|
enQuireBio™
QP8177-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043091
|
enQuireBio™
QP8177-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_065239.1 | |
Serum response factor Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
20807 | |
Research Use Only | |
Srf | |
Recombinant Protein | |
E. coli | |
MLPSQAGAAA ALGRGSALGG NLNRTPTGRP GGGGGTRGAN GGRVPGNGAG LGQSRLEREA AAAAAPTAGA LYSGSEGDSE SGEEEELGAE RRGLKRSLSE MELGVVVGGP EAAAAAAGGY GPVSGAVSGA KPGKKTRGRV KIKMEFIDNK LRRYTTFSKR KTGIMKKAYE LSTLTGTQVL LLVASETGHV YTFATRKLQP MITSETGKAL IQTCLNSPDS PPRSDPTTDQ RMSATGFEEP DLTYQVSESD SSGETKDTLK PAFTVTNLPG TTSTIQTAPS TSTTMQVSSG PSFPITNYLA PVSASVSPSA VSSANGTVLK STGSGPVSSG GLMQLPTSFT LMPGGAVAQQ VPVQAIHVHQ APQQASPSRD SSTDLTQTSS SGTVTLPATI MTSSVPTTVG GHMMYPSPHA VMYAPTSGLA DGSLTVLNAF SQAPSTMQVS HSQVQEPGGV PQVFLTAPSG TVQIPVSAVQ LHQMAVIGQQ AGSSSNLTEL QVVNLDATHS TKSE | |
Greater than 90% as determined by SDS-PAGE. |