missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serum response factor Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum response factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8177-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065239.1 | |
Serum response factor Protein | |
Research Use Only | |
Srf | |
Recombinant Protein | |
E. coli | |
MLPSQAGAAA ALGRGSALGG NLNRTPTGRP GGGGGTRGAN GGRVPGNGAG LGQSRLEREA AAAAAPTAGA LYSGSEGDSE SGEEEELGAE RRGLKRSLSE MELGVVVGGP EAAAAAAGGY GPVSGAVSGA KPGKKTRGRV KIKMEFIDNK LRRYTTFSKR KTGIMKKAYE LSTLTGTQVL LLVASETGHV YTFATRKLQP MITSETGKAL IQTCLNSPDS PPRSDPTTDQ RMSATGFEEP DLTYQVSESD SSGETKDTLK PAFTVTNLPG TTSTIQTAPS TSTTMQVSSG PSFPITNYLA PVSASVSPSA VSSANGTVLK STGSGPVSSG GLMQLPTSFT LMPGGAVAQQ VPVQAIHVHQ APQQASPSRD SSTDLTQTSS SGTVTLPATI MTSSVPTTVG GHMMYPSPHA VMYAPTSGLA DGSLTVLNAF SQAPSTMQVS HSQVQEPGGV PQVFLTAPSG TVQIPVSAVQ LHQMAVIGQQ AGSSSNLTEL QVVNLDATHS TKSE | |
Greater than 90% as determined by SDS-PAGE. |
20807 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |