missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-2 (Cys160Ser) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£221.38
Specifications
Gene ID (Entrez) | 16183 |
---|---|
Molecular Weight (g/mol) | 17 kDa |
Name | IL-2 (Cys160Ser) |
Quantity | 20μg |
Source | E. coli-derived mouse IL-2 protein Ala21-Gln169 (Cys160Ser),with and without an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18136590
|
R&D Systems™
1150-ML-020 |
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. |
£221.38
20µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18128220
|
R&D Systems™
1150-ML-020/CF |
Lyophilized from a 0.2μm filtered solution in Sodium Acetate. |
£221.38
20µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse IL-2 (Cys160Ser) Protein is derived from E. coli. The Recombinant Mouse IL-2 (Cys160Ser) Protein has been validated for the following applications: Bioactivity.Specifications
16183 | |
IL-2 (Cys160Ser) | |
E. coli-derived mouse IL-2 protein Ala21-Gln169 (Cys160Ser),with and without an N-terminal Met | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |
17 kDa | |
20μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |