missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-2 (Cys160Ser) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1150-ML-020/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18128220

  • £221.38 / 20µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse IL-2 (Cys160Ser) Protein is derived from E. coli. The Recombinant Mouse IL-2 (Cys160Ser) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
17 kDa
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
16183
IL-2 (Cys160Ser)
E. coli-derived mouse IL-2 protein Ala21-Gln169 (Cys160Ser),with and without an N-terminal Met
<0.1 EU per 1μg of the protein by the LAL method.
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers