missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-2 (Cys160Ser) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1150-ML-020
Additional Details : Weight : 0.00970kg
Description
The Recombinant Mouse IL-2 (Cys160Ser) Protein is derived from E. coli. The Recombinant Mouse IL-2 (Cys160Ser) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. | |
17 kDa | |
20μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
16183 | |
IL-2 (Cys160Ser) | |
E. coli-derived mouse IL-2 protein Ala21-Gln169 (Cys160Ser),with and without an N-terminal Met | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |