missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse GITR Ligand/TNFSF18 (aa 47-173)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£377.96
Specifications
Accession Number | AAO89011 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 8995 |
Molecular Weight (g/mol) | M.W. (Observed): 16-26 kDa, reducing conditions; M.W. (theoretical): 16 kDa |
Name | GITR Ligand/TNFSF18 aa 47-173 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18198460
|
R&D Systems™
2177-GL-025 |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
£377.96
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18166760
|
R&D Systems™
2177-GL-025/CF |
Lyophilized from a 0.2 μm filtered solution in PBS. |
£377.96
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Measured by its ability to stimulate mouse T cell proliferation in the presence of anti-CD3. The ED50 for this effect is 0.02-0.3 μg/mL in the presence of 10 μg/mL of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ).Specifications
AAO89011 | |
8995 | |
GITR Ligand/TNFSF18 aa 47-173 | |
Mouse myeloma cell line, NS0-derived mouse GITR Ligand/TNFSF18 protein Thr47-Ser173, with an N-terminal 10-His tag | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Tnfsf18 | |
Recombinant | |
GITR Ligand/TNFSF18 |
Bioactivity | |
M.W. (Observed): 16-26 kDa, reducing conditions; M.W. (theoretical): 16 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |