missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse GITR Ligand/TNFSF18 (aa 47-173)

Product Code. 18198460
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18198460

Brand: R&D Systems™ 2177GL025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to stimulate mouse T cell proliferation in the presence of anti-CD3. The ED50 for this effect is 0.02-0.3 μg/mL in the presence of 10 μg/mL of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAO89011
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 8995
Molecular Weight (g/mol) M.W. (Observed): 16-26 kDa, reducing conditions; M.W. (theoretical): 16 kDa
Name GITR Ligand/TNFSF18 aa 47-173
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived mouse GITR Ligand/TNFSF18 protein Thr47-Ser173, with an N-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbol Tnfsf18
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein GITR Ligand/TNFSF18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.