missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
£439.00 - £2645.00
Specifications
Name | GH Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Recombinant Mouse Growth Hormone is fully biologically active when compared to standard Human Growth Hormone which is 3 units/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15924488
|
enQuireBio™
QP10638-100UG |
100 μg |
£439.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944488
|
enQuireBio™
QP10638-200UG |
200 μg |
£660.00
200µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934488
|
enQuireBio™
QP10638-1MG |
1 mg |
£2645.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GH Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MATDSRTSWL LTVSLLCLLW PQEASAFPAM PLSSLFSNAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKE EAQQRTDMEL LRFSLLLIQS WLGPVQFLSR IFTNSLMFGT SDRVYEKLKD LEEGIQALMQ ELEDGSPRVG QILKQTYDKF DANMRSDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFV ESSCAF | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Mouse Growth Hormone is fully biologically active when compared to standard Human Growth Hormone which is 3 units/mg. | |
Mouse | |
Untagged | |
Mouse Growth Hormone Recombinant contains 50mM Tris-HCl, pH 8.0, 150mM NaCl buffer. |