missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10638-100ug
Additional Details : Weight : 0.01000kg
Specifications
GH Protein | |
Research Use Only | |
Recombinant Mouse Growth Hormone is fully biologically active when compared to standard Human Growth Hormone which is 3 units/mg. | |
Mouse | |
Untagged | |
Mouse Growth Hormone Recombinant contains 50mM Tris-HCl, pH 8.0, 150mM NaCl buffer. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MATDSRTSWL LTVSLLCLLW PQEASAFPAM PLSSLFSNAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKE EAQQRTDMEL LRFSLLLIQS WLGPVQFLSR IFTNSLMFGT SDRVYEKLKD LEEGIQALMQ ELEDGSPRVG QILKQTYDKF DANMRSDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFV ESSCAF | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |