missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GH Protein

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10638-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934488

  • £2645.00 / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GH Protein
Research Use Only
Recombinant Mouse Growth Hormone is fully biologically active when compared to standard Human Growth Hormone which is 3 units/mg.
Mouse
Untagged
Mouse Growth Hormone Recombinant contains 50mM Tris-HCl, pH 8.0, 150mM NaCl buffer.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MATDSRTSWL LTVSLLCLLW PQEASAFPAM PLSSLFSNAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKE EAQQRTDMEL LRFSLLLIQS WLGPVQFLSR IFTNSLMFGT SDRVYEKLKD LEEGIQALMQ ELEDGSPRVG QILKQTYDKF DANMRSDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFV ESSCAF
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers