missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse BAG family molecular chaperone regulator 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) BAG family molecular chaperone regulator 3, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_038891.4 |
---|---|
Gene ID (Entrez) | 29810 |
Name | BAG family molecular chaperone regulator 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082491
|
enQuireBio™
QP8071-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022501
|
enQuireBio™
QP8071-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092491
|
enQuireBio™
QP8071-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012501
|
enQuireBio™
QP8071-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032501
|
enQuireBio™
QP8071-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002501
|
enQuireBio™
QP8071-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_038891.4 | |
BAG family molecular chaperone regulator 3 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
29810 | |
Research Use Only | |
Bag3 | |
Recombinant Protein | |
Yeast | |
SAATQSPMMQ MASGNGASDR DPLPPGWEIK IDPQTGWPFF VDHNSRTTTW NDPRVPPEGP KDTASSANGP SRDGSRLLPI REGHPIYPQL RPGYIPIPVL HEGSENRQPH LFHAYSQPGV QRFRTEAAAA TPQRSQSPLR GGMTEAAQTD KQCGQMPATA TTAAAQPPTA HGPERSQSPA ASDCSSSSSS ASLPSSGRSS LGSHQLPRGY IPIPVIHEQN ITRPAAQPSF HQAQKTHYPA QQGEYQPQQP VYHKIQGDDW EPRPLRAASP FRSPVRGASS REGSPARSGT PVHCPSPIRV HTVVDRPQPM THREPPPVTQ PENKPESKPG PAGPDLPPGH IPIQVIRREA DSKPVSQKSP PPAEKVEVKV SSAPIPCPSP SPAPSAVPSP PKNVAAEQKA APSPAPAEPA APKSGEAETP PKHPGVLKVE AILEKVQGLE QAVDSFEGKK TDKKYLMIEE YLTKELLALD SVDPEGRADV RQARRDGVRK VQTILEKLEQ KAIDVPGQVQ VYELQPSNLE AEQPLQEIMG AVVADKDKKG PENKDPQTES QQLEAKAATP PNPSNPADSA GNLVAP | |
Greater than 90% as determined by SDS-PAGE. |